Skip to Content
Merck

D6000

2′-Deoxyadenosine 5′-di­phos­phate sodium salt

≥95% (HPLC), synthetic (organic), powder

Synonym(s):

dADP

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C10H15N5O9P2 · xNa+
CAS Number:
Molecular Weight:
411.20 (free acid basis)
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
276-280-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

2′-Deoxyadenosine 5′-di­phos­phate sodium salt,

InChI key

KZGAPWRJMWSNQO-UHFFFAOYSA-N

InChI

1S/C10H15N5O9P2.Na.H/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(23-7)2-22-26(20,21)24-25(17,18)19;;/h3-7,16H,1-2H2,(H,20,21)(H2,11,12,13)(H2,17,18,19);;

SMILES string

[Na].Nc1ncnc2n(cnc12)C3CC(O)C(COP(O)(=O)OP(O)(O)=O)O3

biological source

synthetic (organic)

assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

Quality Level

General description

2′-Deoxyadenosine5′-di­phos­phate (dADP) is a purine containing adenine as the nucleobase. It isproduced as a product of hypoxanthine oxidase action on hypoxanthine duringpurine degradation in the body.

Application

2′-Deoxyadenosine 5′-diphosphate (dADP) may be used for the synthesis of deoxyadenylate oligonucleotides with enzymes such as polynucleotide phosphorylase from Escherichia coli. dADP is used as an inhibitor of bacterial poly(A) polymerase. DeoxyADP may be use as an alternative substrate or inhibitor for studies of ATPase and DNA and RNA polymerase specificity.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Deutscher
The Journal of biological chemistry, 253(16), 5579-5584 (1978-08-25)
Poly(A) synthesis and degradation have been examined in Escherichia coli cells made permeable to nucleotides by treatment with toluene. Although newly synthesized poly(A) is normally rapidly degraded in this system, extraction of the soluble portion of the cell effectively eliminates
Kevin N Kirouac et al.
Journal of molecular biology, 407(3), 382-390 (2011-02-08)
The ability of DNA polymerases to differentiate between ribonucleotides and deoxribonucleotides is fundamental to the accurate replication and maintenance of an organism's genome. The active sites of Y-family DNA polymerases are highly solvent accessible, yet these enzymes still maintain a
S Gillam et al.
Nucleic acids research, 2(5), 613-624 (1975-05-05)
Polynucleotide phosphorylase from Escherichia coli can be used to catalyse the addition of short tracts of deoxyadenylate residues to the 3'-termini of deoxyribooligonucleotides of the type pdAn-dN (where dN = dC, dT or dG) using dADP as donor. Similarly, the
Mahmoud Kandeel et al.
Journal of bioenergetics and biomembranes, 42(5), 361-369 (2010-08-17)
Nucleoside diphosphate kinases (NDKs) play a key role in maintaining the intracellular energy resources as well as the balance of nucleotide pools. Recently, attention has been directed to NDKs owing to its role in activating various chemotherapeutic agents. The binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service