Skip to Content
Merck
All Photos(1)

Key Documents

D5546

Sigma-Aldrich

DMEM - low glucose

With sodium bicarbonate and sodium pyruvate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: yes
phenol red: yes
glucose: low
HEPES: no
sodium pyruvate: yes
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains a higher concentration of amino acids, vitamins, and additional supplementary components. The original DMEM formula, first reported for culturing embryonic mouse cells, contained 1,000 mg/L of glucose. An alteration with 4,500 mg/L glucose is optimal in cultivating certain cell types. This modification has been shown to improve the stability of the medium. 

Application

Dulbecco′s Modified Eagle′s Medium - low glucose has been used to culture:

  • human osteosarcoma cells and HeLa cells
  • primary human mesenchymal stromal cells (hBMSCs)
  • human embryonic kidney cell lines

Reconstitution

Supplement with 0.584 g/L glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratap C Naha et al.
Toxicology and applied pharmacology, 246(1-2), 91-99 (2010-04-28)
The immunotoxicity of three generations of polyamidoamine (PAMAM) dendrimers (G-4, G-5 and G-6) was evaluated in mouse macrophage cells in vitro. Using the Alamar blue and MTT assays, a generation dependent cytotoxicity of the PAMAM dendrimers was found whereby G-6
Phuc Van Pham et al.
Stem cell research & therapy, 4(4), 91-91 (2013-08-07)
Adipose-derived stem cells (ADSCs) have been isolated, expanded, and applied in the treatment of many diseases. ADSCs have also been used to treat injured articular cartilage. However, there is controversy regarding the treatment efficiency. We considered that ADSC transplantation with
Jun Miyaguchi et al.
The Tohoku journal of experimental medicine, 245(4), 269-275 (2018-08-31)
Bacteria have been used for more than a century to treat solid tumors. Because solid tumors generate an anaerobic environment, we evaluated the anti-tumor effect of the obligate anaerobe strain KK378, derived from Lactobacillus casei (L. casei), using mice bearing
Scott A Hinger et al.
Cell reports, 25(3), 715-725 (2018-10-18)
The regulation and functional roles of secreted coding and long noncoding RNAs (lncRNAs; >200 nt) are largely unknown. We previously showed that mutant KRAS colorectal cancer (CRC) cells release extracellular vesicles (EVs) containing distinct proteomes, microRNAs (miRNAs), and circular RNAs.
Lei Xi Chen et al.
The Laryngoscope, 129(9), 2139-2146 (2019-03-19)
Surgical adhesives are increasingly used for vocal fold microsurgery to assist wound closure and reduce the risks of scar formation. Currently used vocal fold adhesives such as fibrin glue, however, have thus far not been found to promote wound closure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service