Skip to Content
Merck
All Photos(1)

Key Documents

D5523

Sigma-Aldrich

Dulbecco′s Modified Eagle′s Medium - low glucose

With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

DME, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

L-glutamine: yes
NaHCO3: no
sodium pyruvate: yes
phenol red: yes
HEPES: no
glucose: low

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s medium (DME) is a modification of Basal Medium Eagle (BME) that comprises a four-fold higher concentration of amino acids and vitamins, as well as additional supplementary components. The original DME formula, first reported for culturing embryonic mouse cells, contained 1,000 mg/L of glucose. An alteration with 4,500 mg/L glucose is optimal in cultivating certain cell types.

Application

Dulbecco′s Modified Eagle′s medium-low glucose has been used in the differentiation and culture of human and murine mesenchymal stem cells (MSCs).

Quantity

Formulated to contain 10.0 grams of powder per liter of medium.

Reconstitution

Supplement with 3.7 g/L sodium bicarbonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Steffens et al.
Journal of biomaterials applications, 32(5), 663-676 (2017-10-11)
Laminin-functionalized poly-d,l-lactic acid scaffolds were produced. Following this, mesenchymal stem cells and keratinocytes were seeded on biomaterials for the in vivo experiments, where the biomaterials with or without cells were implanted. The analysis is comprised of the visual aspect and mean
Gayatri Jamwal et al.
Biochimica et biophysica acta, 1865(6), 920-931 (2018-04-06)
IGF1R is a ubiquitous receptor tyrosine kinase that plays critical roles in cell proliferation, growth and survival. Clinical studies have demonstrated upregulation of IGF1R mediated signaling in a number of malignancies including colon, breast, and lung cancers. Overexpression of the
Akon Higuchi et al.
Scientific reports, 5, 10217-10217 (2015-05-15)
Human adipose-derived stem cells (hADSCs) exhibit heterogeneous characteristics, indicating various genotypes and differentiation abilities. The isolated hADSCs can possess different purity levels and divergent properties depending on the purification methods used. We developed a hybrid-membrane migration method that purifies hADSCs
Ming-Cheng Chen et al.
Environmental toxicology, 33(5), 587-593 (2018-02-13)
Colorectal cancer (CRC) is one of the most common cancers and causes of cancer-related death. There are several first-line chemotherapeutic drugs used to treat CRC. Oxaliplatin (OXA) is an alkylating cytotoxic agent that is usually combined with other chemotherapeutic drugs
Kallyne K Mimura et al.
Molecular vision, 18, 1583-1593 (2012-06-29)
The aim of this study was to evaluate the expression of the protein annexin A1 (ANXA1), a potent endogenous regulator of the inflammatory process, in ocular toxoplasmosis. C57BL/6 female mice were infected using intravitreal injections of either 10(6) tachyzoites of

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service