Skip to Content
Merck
All Photos(3)

Documents

D5287

Sigma-Aldrich

2′-Deoxyinosine

≥98%

Synonym(s):

9-(2-Deoxy-β-D-ribofuranosyl)hypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O4
CAS Number:
Molecular Weight:
252.23
Beilstein:
33517
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

impurities

inosine, essentially free

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H12N4O4/c15-2-6-5(16)1-7(18-6)14-4-13-8-9(14)11-3-12-10(8)17/h3-7,15-16H,1-2H2,(H,11,12,17)/t5-,6+,7+/m0/s1

InChI key

VGONTNSXDCQUGY-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′-Deoxyinosine has been used in the quantification of nucleoside forms of DNA lesion in a single DNA sample by liquid chromatography tandem mass spectrometry (LC-MS/MS). It has also been used as a standard for high performance liquid chromatography analysis.

Biochem/physiol Actions

2′-Deoxyinosine is a nucleoside composed of hypoxanthine attached to 2′-deoxyribose via a β-N9-glycosidic bond. 2′-Deoxyinosine in DNA can arise from deamination of adenosine. 2′-deoxyinsine can be used as a model compound to study the chemistry of adduct formation and radical chemistry that may affect DNA structures. 2′-Deoxyinosine is used to produce hybridization-sensitive fluorescent DNA probes with self-avoidance ability.
2′-Deoxyinosine is a nucleoside form of hypoxanthine. It is a DNA damage product resulting from the impairment of DNA by reactive nitrogen species. 2′-deoxyinosine is formed from nitrosative deamination by N2O3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuji Ikeda et al.
Organic & biomolecular chemistry, 8(3), 546-551 (2010-01-22)
Hybridization-sensitive fluorescent probes have an inherent disadvantage: self-dimerization of the probe prevents the fluorescence quenching prior to hybridization with the target, resulting in a high background signal. To avoid self-dimerization of probes, we focused on a base pair formed by
Bo Pang et al.
Carcinogenesis, 28(8), 1807-1813 (2007-03-10)
In an effort to define the prevalent DNA damage chemistry-associated chronic inflammation, we have quantified 12 DNA damage products in tissues from the SJL mouse model of nitric oxide (NO) overproduction. Using liquid chromatography-mass spectrometry/MS and immunoblot techniques, we analyzed
Quantification of DNA damage products resulting from deamination, oxidation and reaction with products of lipid peroxidation by liquid chromatography isotope dilution tandem mass spectrometry
Taghizadeh K, et al.
Nature Protocols, 3(8), 1287-1287 (2008)
Manabu Yasui et al.
Journal of molecular biology, 377(4), 1015-1023 (2008-02-29)
Chronic inflammation involving constant generation of nitric oxide (*NO) by macrophages has been recognized as a factor related to carcinogenesis. At the site of inflammation, nitrosatively deaminated DNA adducts such as 2'-deoxyinosine (dI) and 2'-deoxyxanthosine are primarily formed by *NO
Rongjuan Mi et al.
Mutation research, 735(1-2), 12-18 (2012-06-06)
The human endonuclease V gene is located in chromosome 17q25.3 and encodes a 282 amino acid protein that shares about 30% sequence identity with bacterial endonuclease V. This study reports biochemical properties of human endonuclease V with respect to repair

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service