Skip to Content
Merck
All Photos(1)

Key Documents

D3816

Sigma-Aldrich

Anti-Drebrin antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-developmentally-regulated brain protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat

technique(s)

western blot: 1:1,000 using rat brain cytosol extract
western blot: 1:1,000 using whole extract of the human epitheloid carcinoma HeLa cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DBN1(1627)
rat ... Dbn1(81653)

General description

Drebrin is a member of the newly identified ADF-H family of actin-binding proteins that share the structurally conserved actin-depolymerizing factor (ADF) binding module. It is composed of a single ADF-H domain at its N-terminus, followed by a non-conserved central region and a C-terminal SH3 domain. Three isoforms, E1 and E2 (embryonic) and A (adult), have been identified in the brain. An additional truncated form of drebrin A, s-drebrin/A2, is specifically expressed in the adult brain, but not in non-neuronal tissues. Drebrin is also widely expressed in a variety of cells including epithelial, endothelial, and smooth muscle cells and is associated at cell-cell adhering junction sites.

Immunogen

synthetic peptide corresponding to the N-terminal region of human drebrin E/A (amino acids 22-42) conjugated to KLH.

Biochem/physiol Actions

Drebrin (developmentally-regulated brain protein) is a major neuronal F-actin binding protein involved in the control of actin dynamics and neuronal morphogenesis. It colocalizes with actin filaments and dendritic-like cell processes and may be involved in actin remodeling. It binds to and dissociates F-actin stabilizing proteins such as α-actinin, fascin, and tropomyosin from actin filaments. It plays an important role in the formation of axons and dendrites during neuronal development and in neuronal plasticity in the adult brain.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Ishikawa et al.
The Journal of biological chemistry, 269(47), 29928-29933 (1994-11-25)
Drebrin is a development-associated neuroprotein whose cDNA into fibroblasts causes the formation of dendrite-like structures (Shirao, T., Kojima, N., and Obata, K. (1992) Neuroreport 3, 109-112). To explore molecular functions of drebrin during brain development, we purified drebrin from brains
H Asada et al.
Journal of neuroscience research, 38(2), 149-159 (1994-06-01)
Drebrins are developmentally regulated actin-binding proteins. In this study, we analyzed subcellular distribution of drebrin E in neuroblastoma cells (SH-SY5Y) in culture, especially in terms of its relationship to actin filaments. In undifferentiated cells, drebrin E was scattered as flocculus
The ADF homology (ADF-H) domain: a highly exploited actin-binding module.
P Lappalainen et al.
Molecular biology of the cell, 9(8), 1951-1959 (1998-08-07)
T Shirao et al.
Experimental cell research, 215(1), 145-153 (1994-11-01)
Drebrin A is a neuron-specific protein, the expression of which is regulated during development. Upon transfection of fibroblasts with drebrin A cDNA, the protein is expressed at high levels in fibroblasts and the outgrowth of highly branched, neurite-like cell processes
Y Sasaki et al.
Journal of neurochemistry, 66(3), 980-988 (1996-03-01)
The purification of drebrin, an actin-binding protein that is specifically expressed in embryonic rat brain, was described previously. During the purification of drebrin, we found that an actin-binding protein of 54 kDa was also expressed at high levels in embryonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service