Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H12N6O4S
CAS Number:
Molecular Weight:
288.28
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
EC Number:
230-808-0
NACRES:
NA.85
form
powder
Quality Segment
solubility
DMSO: 1 mg/mL
mode of action
cell membrane | interferes
storage temp.
−20°C
SMILES string
OS(O)(=O)=O.NNC1=NNC(=N/N)\c2ccccc12
InChI
1S/C8H10N6.H2O4S/c9-11-7-5-3-1-2-4-6(5)8(12-10)14-13-7;1-5(2,3)4/h1-4H,9-10H2,(H,11,13)(H,12,14);(H2,1,2,3,4)
InChI key
BWHAMWGGORIDBK-UHFFFAOYSA-N
Biochem/physiol Actions
Dihydralazine sulfate is a vasodilator and an antihypertensive agent. It relaxes arterial smooth muscle by inhibiting the accumulation of intracellular free calcium. By relaxing vascular smooth muscle, vasodilators act to decrease peripheral resistance.
Other Notes
Keep container tightly closed in a dry and well-ventilated place.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.