Skip to Content
Merck

D2672

Nalgene® desiccator

overall I.D. × H 150 mm × 150 mm

Synonym(s):

autoclavable polypropylene desiccator, desicctor without vacuum, Nalgene 5315-0150, desiccator, laboratory desiccator

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.43
UNSPSC Code:
41104510
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 1 ea

manufacturer/tradename

Nalgene® 5315-0150

I.D. × H

150 mm × 150 mm

General description

Blue polypropylene body; clear polycarbonate cover. Autoclavable.

Other Notes

Supplied with 140 mm aluminum plate.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library