Skip to Content
Merck

D224

4,5-Diaminofluorescein

5 mM in DMSO, ≥97% (HPLC)

Synonym(s):

DAF-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116100
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solution

availability

not available in Japan

concentration

5 mM in DMSO

storage temp.

2-8°C

Application

Fluorescent detector of nitric oxide in culture medium, tissue sections and biopsy cells.

Analysis Note

Final solution contains approx. 10% isopropanol by volume.

Legal Information

DAF-2 is manufactured by Daiichi Pure Chemicals Co., LTD.
US Patent No. 5,874,590


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Georgina A Rivera-Ingraham et al.
Frontiers in zoology, 13, 4-4 (2016-02-05)
Reactive oxygen (ROS) and nitrogen (RNS) species are produced during normal unstressed metabolic activity in aerobic tissues. Most analytical work uses tissue homogenates, and lacks spatial information on the tissue specific sites of actual ROS formation. Live-imaging techniques (LIT) utilize
Julia Strahl et al.
PloS one, 15(5), e0232360-e0232360 (2020-05-08)
The free radical nitric oxide (NO) is a powerful metabolic regulator in vertebrates and invertebrates. At cellular concentrations in the nanomolar range, and simultaneously reduced internal oxygen partial pressures (pO2), NO completely inhibits cytochrome-c-oxidase (CytOx) activity and hence mitochondrial- and
Isabella Echeverri et al.
American journal of human biology : the official journal of the Human Biology Council, 27(6), 822-831 (2015-05-07)
To evaluate the Relationship between maternal and newborn endothelial function and oxidative stress. Forty-three pregnant women and their offspring were evaluated. As markers of endothelial function, the flow-mediated dilation (FMD) was measured in pregnant women in the second and third



Global Trade Item Number

SKUGTIN
D224-1MG04061833561980