Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C19H26I3N3O9
CAS Number:
Molecular Weight:
821.14
UNSPSC Code:
12352207
NACRES:
NA.75
PubChem Substance ID:
EC Number:
266-164-2
Beilstein/REAXYS Number:
2406632
MDL number:
Quality Segment
form
powder
application(s)
hematology
histology
storage temp.
room temp
SMILES string
CC(=O)N(CC(O)CO)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I
InChI
1S/C19H26I3N3O9/c1-8(29)25(4-11(32)7-28)17-15(21)12(18(33)23-2-9(30)5-26)14(20)13(16(17)22)19(34)24-3-10(31)6-27/h9-11,26-28,30-32H,2-7H2,1H3,(H,23,33)(H,24,34)
InChI key
NTHXOOBQLCIOLC-UHFFFAOYSA-N
General description
HistoDenz™, also known as iohexol, is a non-ionic X-ray contrast compound developed specifically as an iodinated density gradient medium.
Application
HistoDenz™ is suitable for separating everything from macromolecules to viable cells by either rate-zonal or isopycnic centrifugation.
- It has been used in the isolation and culturing of hepatic stellate cells.
- It has also been employed in the extraction of microbial cells from environmental samples.
- It has been used in LacZ (β-galactosidase) staining.
- It has also been used for the enrichment of gonocytes.
Features and Benefits
HistoDenz™ offers several favorable properties as a gradient medium.
- It has very low toxicity, low osmolality, high density, and low viscosity (compared to sucrose).
- It is water-soluble, non-ionic, and chemically near-inert.
Legal Information
Histodenz is a trademark of Sigma-Aldrich Co. LLC
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Isolation and Culture of Hepatic Stellate Cells
Ralf Weiskirchen, Axel M. Gressner
Methods in Molecular Medicine, 117 (2005)
Extraction of Microbial Cells from Environmental Samples for FISH Approaches
Jennifer Pratscher
Methods in Molecular Biology, 2246 (2021)
Attachment of Pseudomonas putida onto differently structured kaolinite minerals: A combined ATR-FTIR and 1H NMR study.
Vasiliadou I A, et al.
Colloids and Surfaces. B, Biointerfaces, 84(2), 354-359 (2011)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D2158-100G | 04061833561768 |