Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

Merck

Key Documents

Skip To

D1782

1,2-Dioleoyl-3-palmitoyl-rac-glycerol

≥99% (TLC)

Synonym(s):

1,2-Di(cis-9-octadecenoyl)-3-hexadecanoyl-rac-glycerol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C55H102O6
CAS Number:
Molecular Weight:
859.39
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥99% (TLC)

form

liquid

functional group

ketone

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\CCCCCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C55H102O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h25-28,52H,4-24,29-51H2,1-3H3/b27-25+,28-26+

InChI key

JFISYPWOVQNHLS-NBHCHVEOSA-N

Packaging

Sealed ampule

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0301D2135D3627
description

≥99% (TLC)

description

≥95% (TLC), liquid

description

≥99%

description

≥99% (GC)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

plant

assay

≥99% (TLC)

assay

≥95% (TLC)

assay

≥99%

assay

≥99% (GC)

functional group

ketone

functional group

ester

functional group

ester

functional group

oleic acid

form

liquid

form

liquid

form

powder

form

-

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters