Skip to Content
Merck
All Photos(4)

Key Documents

D1634

Sigma-Aldrich

Diosgenin

≥93%

Synonym(s):

(25R)-5-Spirosten-3β-ol, 3β-Hydroxy-5-spirostene, Nitogenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42O3
CAS Number:
Molecular Weight:
414.62
Beilstein:
94582
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥93%

form

powder

SMILES string

[H][C@]12C[C@@]3([H])[C@]4([H])CC=C5C[C@@H](O)CC[C@]5(C)[C@@]4([H])CC[C@]3(C)[C@@]1([H])[C@H](C)[C@@]6(CC[C@@H](C)CO6)O2

InChI

1S/C27H42O3/c1-16-7-12-27(29-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(28)8-10-25(18,3)21(20)9-11-26(22,24)4/h5,16-17,19-24,28H,6-15H2,1-4H3/t16-,17+,19+,20-,21+,22+,23+,24+,25+,26+,27-/m1/s1

InChI key

WQLVFSAGQJTQCK-VKROHFNGSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

Diosgenin is a triterpene and a glycone derivative of dioscin. Yams, Trigonella foenum-graecum and Costus speciosus are some of its source.

Application

Diosgenin has been:
  • used as a standard in the analysis of total saponins in Sapindus rarak
  • orally administered to the rats to study its role against atherogenic diet-induced production of inflammatory mediators in heart, liver and brain
  • used as a standard compound to determine the saponins content in chickpea and Tribulus terrestris

Biochem/physiol Actions

Diosgenin induces apoptosis in colon cancer cell lines and induces apoptosis, cell cycle arrest and cyclooxygenase activity in osteosarcoma cells. It serves as a precursor in steroid drug production. Diosgenin is shown to promote cholesterol production by stimulating biliary excretion. It influences lipoxygenase induced human erythroleukemia cell line differentiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaofang Hua et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 1460-1465 (2016-11-05)
Gestational diabetes (GD) is a pathological condition, affecting 2-5% of pregnant women. Diosgenin (DSG) possesses a variety of biological activities. The present study was designed to examine the effect of DSG on GD and to investigate the possible mechanism in
Extraction of tannins and saponins from plant sources and their effects on in vitro methanogenesis and rumen fermentation
Yuliana P, et al.
Journal of the Indonesian Tropical Animal Agriculture, 39(2), 91-97 (2014)
Jing Chen et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 36(6), 806-810 (2016-12-08)
This study investigated the effect of diosgenin, a natural sapogenin possessing various pharmacological activities, on benign prostatic hyperplasia (BPH) in rats and the possible mechanisms. BPH was established in the castrated rats by subcutaneous injection of testosterone propionate. Animals were
Lijuan Wang et al.
Journal of natural products, 76(5), 909-914 (2013-04-30)
The purpose of this study was to investigate the ameliorating effect of dioscin (1) on multidrug resistance (MDR) in adriamycin (ADR)-resistant erythroleukemic cells (K562/adriamycin, K562/ADR) and to clarify the molecular mechanisms involved. High levels of multidrug resistance 1 (MDR1) mRNA
Diosgenin induces cell cycle arrest and apoptosis in human leukemia K562 cells with the disruption of Ca 2+ homeostasis
Liu MJ, et al.
Cancer Chemotherapy and Pharmacology, 55(1), 79-90 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service