Skip to Content
Merck

D1414

Droperidol

dopamine subtype 2 receptor antagonist, powder

Synonym(s):

1-[1-[3-(p-Fluorobenzoyl)propyl]-1,2,3,6-tetrahydro-4-pyridyl]-2-benzimidazolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H22FN3O2
CAS Number:
Molecular Weight:
379.43
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
208-957-8
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Droperidol,

form

powder

Quality Segment

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCC(=CC2)N3C(=O)Nc4ccccc34

InChI

1S/C22H22FN3O2/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-11H,3,6,12-15H2,(H,24,28)

InChI key

RMEDXOLNCUSCGS-UHFFFAOYSA-N

Biochem/physiol Actions

Droperidol is a potent antagonist of dopamine subtype 2 receptor. It is an antipsychotic, which is used to treat acute behavioral anomalies. It helps to manage postoperative nausea and vomiting.
D1, D2 dopamine receptor antagonist; butyrophenone antipsychotic and anti-emetic.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Droperidol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library