Skip to Content
Merck
All Photos(2)

Documents

D0540

Sigma-Aldrich

DNQX

≥98% (TLC)

Synonym(s):

6,7-Dinitroquinoxaline-2,3(1H,4H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4N4O6
CAS Number:
Molecular Weight:
252.14
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

SMILES string

[O-][N+](=O)c1cc2NC(=O)C(=O)Nc2cc1[N+]([O-])=O

InChI

1S/C8H4N4O6/c13-7-8(14)10-4-2-6(12(17)18)5(11(15)16)1-3(4)9-7/h1-2H,(H,9,13)(H,10,14)

InChI key

RWVIMCIPOAXUDG-UHFFFAOYSA-N

Application

DNQX has been used to block N-Methyl-D-aspartic acid (NMDA) and α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptors in various experiments.

Biochem/physiol Actions

DNQX is a quinoxaline derivative.
A competitive kainate, quisqualate (non-NMDA) glutamate receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Focal activation of neuronal circuits induced by microstimulation in the visual cortex
Tanaka Y, et al.
Journal of Neural Engineering, 16(3), 036007-036007 (2019)
Rei Yamada et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(9), 3927-3938 (2013-03-01)
Neurons in the nucleus laminaris (NL) of birds detect the coincidence of binaural excitatory inputs from the nucleus magnocellularis (NM) on both sides and process the interaural time differences (ITDs) for sound localization. Sustained inhibition from the superior olivary nucleus
The mouse pulvinar nucleus links the lateral extrastriate cortex, striatum, and amygdala
Zhou N, et al.
The Journal of Neuroscience, 38(2), 347-362 (2018)
James M Otis et al.
Neuron, 103(3), 423-431 (2019-06-15)
The paraventricular thalamus (PVT) is an interface for brain reward circuits, with input signals arising from structures, such as prefrontal cortex and hypothalamus, that are broadcast to downstream limbic targets. However, the precise synaptic connectivity, activity, and function of PVT circuitry
Julian M C Choy et al.
Cerebral cortex (New York, N.Y. : 1991), 27(1), 589-601 (2015-10-28)
Despite its comparatively simple trilaminar architecture, the primary olfactory (piriform) cortex of mammals is capable of performing sophisticated sensory processing, an ability that is thought to depend critically on its extensive associational (intracortical) excitatory circuits. Here, we used a novel

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service