Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Conjugate:
unconjugated
Clone:
DDC-109, monoclonal
Application:
ARR, ELISA (i), ICC, IP, WB
Citations:
4
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
DDC-109, monoclonal
mol wt
antigen ~50 kDa
contains
15 mM sodium azide
species reactivity
rabbit, sheep, guinea pig, monkey, rat, human, bovine, canine
technique(s)
immunocytochemistry: suitable, immunoprecipitation (IP): suitable, indirect ELISA: suitable, microarray: suitable, western blot: 1:1,000 using bovine brain extract
isotype
IgG2b
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... DDC(1644)
rat ... Ddc(24311)
General description
DDC is a 85-90 kDa molecular weight protein that is mainly expressed in brain, kidney and liver.
Monoclonal Anti-DOPA Decarboxylase (DDC) (mouse IgG2b isotype) is derived from the DDC-109 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
Application
Monoclonal Anti-DOPA Decarboxylase (DDC) antibody produced in mouse may also be used in immunoprecipitation, immunocytochemistry and ELISA applications. In immunocytochemical labeling of brain tissue, the product stains the monoaminergic cells and proximal dendrites, but not cortical fibers. It is suitable for use in western blotting at a working dilution of 1:1000 using bovine brain extract. It was used for immunoblotting and immunoprecipitation of AADC (aromatic amino acid decarboxylase) from rat striatum in a study. It was used as a primary antibody for western blotting and co-immunoprecipitation studies using human embryonic kidney cells and human neuroblastoma cells.
Biochem/physiol Actions
DOPA Decarboxylase (DDC), which requires pyridoxal phosphate for its activity, decarboxylates 5-hydroxytryptophan to form serotonin. It also decarboxylates L-DOPA to form dopamine in both dopaminergic and noradrenergic neurons.
DOPA decarboxylase (DDC, EC 4.1.1.28), catalyzes the decarboxylation of the 5-hydroxytrptophan (5-HTP) to 5-HT. It requires pyridoxal phosphate for its activity. This is the enzyme that also decarboxylates L-DOPA to form dopamine in both dopaminergic and noradrenergic.. Antibodies that react specifically with the enzyme enable the identification of both noradrenergic, dopaminergic, and serotoninergic systems, since DDC is present and active in all three types of neurons.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D0180-.2ML | 04061837720079 |