Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352207
EC Number:
217-896-6
NACRES:
NA.32
Technique(s):
flow cytometry: suitable, protein staining: suitable
Usage:
kit sufficient for 1000 reactions (using 96 multiwell plates), kit sufficient for 50 reactions (in a 2 mL cuvette)
Storage temp.:
−20°C
Detection method:
fluorometric
Shipped in:
dry ice
Quality Segment
usage
kit sufficient for 1000 reactions (using 96 multiwell plates), kit sufficient for 50 reactions (in a 2 mL cuvette)
packaging
pkg of 1 kit
storage condition
dry at room temperature
technique(s)
flow cytometry: suitable, protein staining: suitable
fluorescence
λex 490 nm; λem 590 nm (JC-1 dye)
application(s)
cell analysis
detection
detection method
fluorometric
shipped in
dry ice
storage temp.
−20°C
Application
Isolated Mitochondria Staining Kit has been used to assess mitochondrial inner membrane potential.
Biochem/physiol Actions
The kit is based on mitochondria staining using the JC-1 dye. In normal cells, the JC-1 dye concentrates in the mitochondrial matrix where it forms red fluorescent aggregates. Any event that dissipates the mitochondrial membrane potential (e.g. apoptosis) does not allow the accumulation of JC-1 dye in the mitochondria. The dye is detected using a fluorimeter or fluorescence microscope.
Features and Benefits
- Fast, simple, and convenient method for staining isolated mitochondria and for assaying mitochondria intactness
- Contains all the reagents required for the detection of changes in mitochondrial inner-membrane electrochemical potential
- Contains the antibiotic Valinomycin that permeabilizes the mitochondrial membrane, and thus, dissipates the mitochondrial electrochemical potential. Valinomycin can be used as a control agent that prevents JC-1 aggregation
Analysis Note
The kit measures mitochondria intactness and detects events that dissipate the mitochondrial membrane potential.
Kit Components Only
Product No.
Description
- DMSO 1 mL
- JC-1 Assay Buffer 5X 25 mL
- JC-1 Stain 25 μg
- Valinomycin Ready Made 100 μL
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 1 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
flash_point_f
Not applicable
flash_point_c
Not applicable
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Subcellular fractionation of mitochondria.
Rice, J.E., and Lindsay, J.G. et al.
Subcellular Fractionation, A Practical Approach, 107- 142 (1997)
Arpan Kumar Maiti et al.
Interdisciplinary toxicology, 11(4), 306-315 (2019-11-26)
Nickel is a potential neurotoxic pollutant inflicting damage in living organisms, including fish, mainly through oxidative stress. Previous studies have demonstrated the impact of nickel toxicity on mitochondrial function, but there remain lacunae on the damage inflicted at mitochondrial respiratory
Differential expression of choline kinase isoforms in skeletal muscle explains the phenotypic variability in the rostrocaudal muscular dystrophy mouse
Wu G, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 446-454 (2010)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| CS0760-1KT | 04061833553138 |
