Skip to Content
Merck

CPS3500

Chemiluminescent Peroxidase Substrate-3

sufficient for 500 mL substrate

Synonym(s):

CPS3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

usage

sufficient for 500 mL substrate

storage temp.

2-8°C

Application

CPS3 formulations are economically priced and outperform the most commonly used competitors′ products. This two component system provides low nanogram detection with superior signal-to-noise ratio. In addition, the components are stable after mixing, adding to the flexibility of usage.


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library