Skip to Content

CPS260

Chemiluminescent Peroxidase Substrate for ELISA

sufficient for 60 mL substrate

Synonym(s):

CPS2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

usage

sufficient for 60 mL substrate

storage temp.

2-8°C

Application

Two component liquid substrate system with superior sensitivity, signal-to-noise, and linearity.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Chemiluminescent Reagent 20 mL
    SDS

  • Chemiluminescent Reaction Buffer 40 mL
    SDS

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library