Skip to Content
Merck

Skip To

CORET

PCR Core kit with Taq DNA polymerase

PCR components in separate tubes to allow for optimization

Synonym(s):

PCR kit, Taq DNA Polymerase kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106300
NACRES:
NA.55
eCl@ss:
32161000
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

usage

sufficient for 250 reactions

feature

dNTPs included, hotstart: no, Standard PCR

technique(s)

PCR: suitable

color

colorless

input

purified DNA

shipped in

wet ice

storage temp.

−20°C

General description

Taq DNA Polymerase is a thermostable enzyme derived from the thermophilic bacterium Thermus aquaticus. This enzyme is in a recombinant form, expressed in E. coli. Taq DNA polymerase enzyme can withstand high temperatures up to 95 °C without significant loss of activity. Taq DNA Polymerase is a magnesium ion-dependent enzyme. Taq DNA Polymerase possesses 5′→3′ DNA polymerase activity and 5′→3′ exonuclease activity.

Application

CORET PCR kit has been used for the amplification of genomic DNA during the study of genetic diversity in various species of Manilkara zapota.
PCR Core kit with Taq DNA polymerase is suitable for the amplification of genomic DNA, cDNA, or cloned DNA templates. Each component has been tested in the polymerase chain reaction. Optimal concentrations of Taq polymerase, template DNA, primers, and MgCl2 will depend on the specifics of the system. It may be necessary to optimize each component separately, especially the Taq and Mg2+ concentrations and the cycling parameters.

Preparation Note

Kit contains enough reagents for 250 units of Taq DNA polymerase.

Other Notes

The following reagents are required but not provided in the kit:
  • Mineral Oil, M 8662 (optional)
  • Thermocycler
  • Primers
  • DNA to be amplified
  • Chloroform, C 7559 (optional)

Legal Information

No license is conveyed with the purchase of this product under any of US Patents Nos. 5,804,375, 5,994,056, 6,171,785, 6,214,979, 5,538,848, 5,723,591, 5,876,930, 6,030,787, and 6,258,569, and corresponding patents outside the United States, or any other patents or patent applications, relating to the 5′ Nuclease and dsDNA-Binding Dye Processes. For further information contact the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D1806D4545D7442
technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

feature

dNTPs included, Standard PCR, hotstart: no

feature

dNTPs included: no, hotstart: no, Standard PCR

feature

dNTPs included: no, hotstart: no, Standard PCR

feature

Difficult Templates/Specialty Enzymes PCR, dNTPs included: no, hotstart: no, Standard PCR

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

input

purified DNA

input

purified DNA

input

purified DNA

input

purified DNA

form

liquid

form

liquid

form

liquid

form

liquid


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • 10X PCR Buffer, Optimized for routine PCR with MgCl2 included 1 mL/vial
    SDS

  • 10X PCR Buffer without MgCl2, Optimized for routine PCR without MgCl2 1 mL/vial
    SDS

  • Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR 1 mL/vial
    SDS

  • Water, PCR Reagent 3 vial(s)
    SDS

  • Deoxynucleotide Mix, 10 mM, Molecular Biology Reagent .25 mL
    SDS

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters