Skip to Content
Merck

CLS8393

Corning® Costar® traditional, round, plastic storage bottles

round clear polystyrene, sterile, bottle capacity 500 mL, cap (plug seal), case of 12

Synonym(s):

plastic bottle, plastic bottles, storage bottle

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.14
UNSPSC Code:
41121800
Material:
cap (plug seal), clear polystyrene bottle, cylindrical bottle, red high-density polyethylene cap (plug seal), round clear polystyrene
Sterility:
sterile
Feature:
cap, graduated, graduations, neck thread wide mouth
Packaging:
case of 12
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cap (plug seal), clear polystyrene bottle, cylindrical bottle, red high-density polyethylene cap (plug seal), round clear polystyrene

sterility

sterile

feature

cap, graduated, graduations, neck thread wide mouth

packaging

case of 12

manufacturer/tradename

Corning 8393

bottle capacity

500 mL

bottle neck diam.

~44 mm

neck diam.

45 mm

General description

These traditional style disposable polystyrene bottles have smooth sides and are ideal for storage of media, buffers and other aqueous solutions. 45 mm polyethylene plug seal cap provides an airtight seal and helps minimize the risk of contamination. Sterile, certified nonpyrogenic.

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library