CLS67814
Corning® LSE™ digital microplate shakers
digital microplate shaker, accommodates up to 4 microplates, 230 V, EU plug, 1/cs
Synonym(s):
laboratory mixer shaker, laboratory shakers, microplate shakers
About This Item
Recommended Products
AC/DC input
230 V AC, Schuko plug
sterility
non-sterile
feature
CE compliant
lid: no
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 6781-4
parameter
100-1400 rpm speed
L × W × H
16.8 cm × 27.9 cm × 15.3 cm
weight
4.3 kg
color
none
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Large capacity - process up to 4 microplates
- 3 mm orbit for resuspending, homogenizing or vortexing
- Large digital display with single-knob control
- Suitable for cold room or incubator use
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service