CLS440129
Support Rod for Corning® hot plates and stirrers
diam. × L 5/16 × 18 in. , two-pieces
All Photos(2)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.73
Recommended Products
sterility
non-sterile
manufacturer/tradename
Corning 440129
diam. × L
5/16 × 18 in. , two-pieces
Related Categories
General description
Corning offers a stainless steel, two piece, 18 in. (45.7cm) support rod with a 5/16 in. (0.8 cm) diameter that consists of two 9 in. rods that screw together. It screws into the base of Corning hot plate and stirrer models PC-400/410/420, PC-400D/410D/420D and PC-600D/610D/620D.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)