CLS431120
Corning® cryogenic boxes, racks & trays
Box for 81 cryogenic vials, 4-5 mL
Synonym(s):
cryovials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123304
NACRES:
NB.13
Recommended Products
sterility
non-sterile
feature
Holds 81 x 4-5 mL tubes
packaging
pack of 10 ea
manufacturer/tradename
Corning 431120
technique(s)
cryopreservation: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Reusable orange cryogenic vial storage box with lid
- Made of virgin polycarbonate to withstand temperature from - 196°C to +121°C
- Ridges on the sides of the box improve grip when wearing gloves
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service