CLS3542
Corning® 384 well microplate, low volume
black polystyrene, flat bottom, sterile, lid, pack of 50
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates
About This Item
clear bottom
clear wells
flat bottom
skirt
Recommended Products
material
black polystyrene
clear bottom
clear wells
flat bottom
sterility
sterile
feature
lid
skirt
packaging
pack of 50
manufacturer/tradename
Corning 3542
size
384 well
well volume
50 μL
well working volume
5-40 μL
wells
384 wells , low volume
suitability
suitable for (molecular, validated for the FLIPR reader)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service