Skip to Content
Merck
All Photos(1)

Key Documents

CLS353144

Corning® Falcon® Cell Culture Multi-Flask

capacity 1750 mL, 5-layer, cap, vented

Synonym(s):

Cell Culture Multi-Flask, Corning Multi-Flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

polystyrene

feature

cap vented

manufacturer/tradename

Corning 353144

technique(s)

cell culture | mammalian: suitable

capacity

1750 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service