CLS353028
Corning® Falcon® Cell Culture Flask
capacity 350 mL, straight neck, graduated, 50 ‑ 600 mL, cap, plug seal
Synonym(s):
Corning container
About This Item
Recommended Products
material
polystyrene
feature
cap plug seal
graduated 50 ‑ 600 mL
manufacturer/tradename
Corning 353028
technique(s)
cell culture | mammalian: suitable
capacity
350 mL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service