CLS3513
Corning® Costar® TC-Treated Multiple Well Plates
size 12 wells, clear polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid
Synonym(s):
cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates
About This Item
clear polystyrene plate
flat bottom
round wells
skirt
Recommended Products
material
clear bottom
clear polystyrene plate
flat bottom
round wells
description
growth area 3.8 cm2
sterility
sterile
feature
lid
skirt
packaging
case of 50 (individually wrapped)
manufacturer/tradename
Corning 3513
size
12 wells
well diam.
22.1 mm
well volume
6.9 mL
well working volume
0.76-1.14 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Corning® Costar® multiple well plates support various assay and storage application requirements.
- TC-treated surface for optimal cell attachment.
- Rings on lid prevent cross-contamination.
- All plates have a uniform footprint and a raised bead to aid stacking.
- Individual alphanumerical codes for well identification.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service