Skip to Content
Merck

CLS3471

Corning® Costar® Ultra-Low Attachment Multiple Well Plate

size 6 wells, flat bottom clear, pkg of (individually wrapped), pkg of (24/case), sterile, lid

Synonym(s):

6 well plate, cell culture plate, tissue culture plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121800
Material:
clear , clear wells, flat bottom clear , polystyrene
Size:
6 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , clear wells, flat bottom clear , polystyrene

sterility

sterile

feature

lid, skirt, plate format: standard, well: flat bottom, clear

manufacturer/tradename

Corning 3471

packaging

pack of 1 (individually wrapped w/ lid), case of 24, pkg of (24/case), pkg of (individually wrapped)

size

6 wells

surface area

9.5 cm2 , cell growth area (cm2)

well volume

16.8 mL

well working volume

1.9-2.9 mL

wells

6 wells

color

clear

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

General description

Corning Costar Ultra-Low Attachment Multiwell Plates

  • Ultra-Low attachment plates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library