Skip to Content
Merck

CHY5S

α-Chymotrypsin from bovine pancreas

≥40 units/mg protein, vial of 5 mg

Synonym(s):

α-chymotrypsin A and B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-671-2
MDL number:
EC Number:
eCl@ss:
42010112
Specific activity:
≥40 units/mg protein
Biological source:
bovine pancreas
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pancreas

Quality Segment

description

aseptically filled

type

Type IV-S

form

solid

specific activity

≥40 units/mg protein

mol wt

25 kDa

composition

protein, ≥85% UV

packaging

vial of 5 mg

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... CTRB1(618826)

Application

α-Chymotrypsin from bovine has been used in a study to inform proteasome inhibition in order to advance anticancer research. α-Chymotrypsin from bovine has also been used in a study that functionalized surface anchored poly(methylhydrosiloxane) thin films on oxidized silicon wafers.
The enzyme from Sigma has been used to assess the effect of limited proteolysis with α-chymotrypsin on the sperm penetration.
α-Chymotrypsin from bovine pancreas has been used:
  • as a supplement for the collection of semen into Tris diluent
  • as one of the proteases in the analysis of major histocompatibility complex (MHC) class II protease sensitivity assay
  • as a component in YEPD broth for biofilm dispersion assay
  • in the preparation of chitinase–chymotrypsin–DMSO buffer (CCD buffer) for enzymatic digestion of larvae

Biochem/physiol Actions

α-Chymotrypsin is a serine peptidase and has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. Molecular weight of this enzyme is found to be 25 kDa. Its pI is 8.75. It selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan, and leucine. Ca2+ activates and stabilizes the enzyme. The enzyme is inhibited by diisopropyl fluorophosphate (DFP), phenylmethanesulfonyl fluoride (PMSF), N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK), chymostatin, aprotinin, α1-antitrypsin, and α2-macroglobulin, 10 mM Cu2+ and Hg2+.
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met, Leu) on the carboxyl end of the bond.

Analysis Note

Protein determined by A1%/280

Other Notes

One unit will hydrolyze 1.0 μmole of BTEE per min at pH 7.8 at 25 °C.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library