Skip to Content
Merck
All Photos(1)

Key Documents

MOR/0.4R

96042334, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

MOR/0.4R, 96042334

biological source

human lung

growth mode

Semi-adherent aggregates

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung adenocarcinoma, drug-resistant

Cell Line Description

MOR/0.4R has been developed from the parent line, MOR, by continuous exposure to increasing concentrations of doxorubicin (also known as adriamycin). This drug-resistant cell line accumulates lower levels of doxorubicin than the parent line and have been shown to overexpress multi drug resistance-associated protein (MRP). The expression of a 190kDa membrane protein associated with the degree of drug resistance has been indicated. Cells grow as easily detaching aggregates

Application

Analysis of acquired drug resistance, MRP studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 12
D16S539: 9
D5S818: 12
D7S820: 9
THO1: 7
TPOX: 11
vWA: 15

Culture Medium

RPMI 1640 + 2mM Glutamine + 0.4μg/ml Doxorubicin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split subconfluent cultures (70-80%) 1:2 to 1:4 seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. It is recommended to culture the cells without drug after resuscitation until the first passage. These cells produce a

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service