Skip to Content
Merck
All Photos(1)

Key Documents

IgH-2

90030804, lizard cardiovascular , Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

IgH-2, 90030804

biological source

lizard cardiovascular

growth mode

Adherent

karyotype

2n = 34

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Iguana heart

Cell Line Description

From the heart of a normal immature male Iguana. Cells can support the replication of herpes simplex, PRV, vaccinia and Iguana viruses at 36°C. The only reported reptilian line to be established and serially propagated at an incubation temperature as high as 36°C.

Application

Virus replication studies: HSV, PRV, vaccinia and Iguana viruses

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 36°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service