Skip to Content
Merck
All Photos(1)

Key Documents

HOS

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87070202, human bone, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human bone

packaging

tube of 5 μg 87070202-DNA-5UG
pkg of vial of cells 87070202-1VL

growth mode

Adherent

karyotype

2n = 46, hyperdiploid

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian osteosarcoma, TE85

Cell Line Description

Derived from a 13 year old female caucasian. Cells are sensitive to both virus and chemical transformation and exhibit a flat morphology, low saturation density and low plating efficiency in soft agar. HOS and 143B (Sigma Catalogue number 91112502) originated from the same host: 143B is TK- and originated indirectly from HOS which is TK+ (Skovgaard et al 2010 PMID: 20544518).

Application

Virus, transformation and biochemical studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 10,13
D5S818: 13
D7S820: 11,12
THO1: 6
TPOX: 8,11
vWA: 18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service