YTH 30.15
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 11060807
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Rat x Rat Hybridoma
packaging
tube of 5 μg 11060807-DNA-5UG
pkg of vial of cells 11060807-1VL
ImmunoDon
DA rat spleen
Myeloma
Y3.AG.1.2.3
Reactivity
Human CD7
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
Looking for similar products? Visit Product Comparison Guide
Cell Line Origin
Monoclonal antibody human CD28
Cell Line Description
Rat hybridoma producing antibody against human CD7. Cambridge Enterprise Reference number AB69:www.enterprise.cam.ac.uk.
Immunogen
Human peripheral blood mononuclear cells
Culture Medium
IDMEM (I3390) + 2 mM L-Glutamine (G7513) + 5% FBS / FCS (F2442). Where the cells are grown under static conditions, 0.02% EDTA, Trypsin (T4424) or mechanical agitation maybe required to dislodge them from the plastic.
Subculture Routine
Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37 °C. Saturation density 9 x 105/ml.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service