Skip to Content
Merck
All Photos(5)

Key Documents

C9758

Sigma-Aldrich

L-Canavanine sulfate salt

≥99% (TLC), powder

Synonym(s):

L-α-Amino-γ-(guanidinooxy)butyric acid sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N4O3 · H2SO4
CAS Number:
Molecular Weight:
274.25
Beilstein:
6121291
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

plant seeds (Jack bean )

Quality Level

Assay

≥99% (TLC)

form

powder

color

white

mp

160-165 °C (dec.)

solubility

H2O: 100 mg/mL

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.N[C@@H](CCONC(N)=N)C(O)=O

InChI

1S/C5H12N4O3.H2O4S/c6-3(4(10)11)1-2-12-9-5(7)8;1-5(2,3)4/h3H,1-2,6H2,(H,10,11)(H4,7,8,9);(H2,1,2,3,4)/t3-;/m0./s1

InChI key

MVIPJKVMOKFIEV-DFWYDOINSA-N

Gene Information

human ... NOS2(4843)

Looking for similar products? Visit Product Comparison Guide

General description

Canavanine is a homolog of arginine and a non-protein amino acid. It is present in plants like alfalfa, cloves, beans onions and sprouts. Canavanine is synthesized from L-canavaninosuccinic acid and the biosynthetic pathway was first elucidated in jack bean plant.

Application

Canavanine sulfate salt has been used:
  • as a translation inhibitor in yeast translation studies
  • as a component of synthetic complete (SC) drop-out medium for screening yeast
  • for screening Saccharomyces cerevisiae in canavanine mutation frequency assay

Biochem/physiol Actions

Canavanine is a naturally occurring L-amino acid that competes and interferes with L-arginine-utilizing enzymes due to its structural similarity with this L-amino acid. It is a selective inhibitor of inducible nitric oxide synthase (iNOS). It acts as a substrate for arginyl tRNA and is incorporated during protein synthesis leading to aberrant canavanyl proteins. Supplementation with canavanine worsens the alfalfa induced Systemic lupus erythematosus (SLE) like syndrome and its incorporation with endosome and proteasome disrupts the antigen processing.
Naturally occurring L-amino acid that interferes with L-arginine-utilizing enzymes due to its structural similarity with this L-amino acid. It is a selective inhibitor of inducible nitric oxide synthase (iNOS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-throughput transformation of Saccharomyces cerevisiae using liquid handling robots
Liu G, et al.
PLoS ONE, 12(3), e0174128-e0174128 (2017)
Toshiya Ogata et al.
Cell biology international, 40(1), 100-106 (2015-10-29)
A mitochondrial superoxide dismutase (SOD2) is the first line of antioxidant defense against mitochondrial superoxide. Even though the involvement of SOD2 in lifespan has been studied extensively in several organisms, characterization of the aging process has not been performed for
Kellyn M Hoffert et al.
Genetics, 213(1), 97-112 (2019-07-20)
Maintenance of genome integrity is a crucial cellular focus that involves a wide variety of proteins functioning in multiple processes. Defects in many different pathways can result in genome instability, a hallmark of cancer. Utilizing a diploid Saccharomyces cerevisiae model
Osama Mansour et al.
Current genetics, 66(3), 577-592 (2020-01-15)
Mutations in budding yeast occur in meiosis at higher frequencies than in cells grown vegetatively. In contrast to mutations that occur in somatic cells, meiotic mutations have a special, long-range impact on evolution, because they are transferred to the following
L-canavanine, a selective inhibitor of inducible NO synthase, increases plasma endothelin-1 concentrations in dogs with endotoxic shock
Mitaka C, et al.
Journal of Critical Care, 16(1), 17-23 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service