Skip to Content
Merck

C9744

CAMK1γ (1-330), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CLICKIII, VWS1, dJ272L16.1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Specific activity:
670-906 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

670-906 nmol/min·mg

mol wt

~69 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAMK1G(57172)

Biochem/physiol Actions

CAMKIγ (CLICK-III), a member of CAMK family, is a novel membrane-anchored neuronal Ca2+/calmodulin-dependent protein kinase. Full activation of CaMKIγ requires both Ca2+/CaM and phosphorylation by CAMKK. CAMKIγ transcripts is most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Sayaka Takemoto-Kimura et al.
The Journal of biological chemistry, 278(20), 18597-18605 (2003-03-15)
During a screen for novel putative Ca(2+)/calmodulin-dependent protein kinase (CaMK)-like CREB kinases (CLICKs), we have cloned a full-length cDNA for CLICK-III/CaMKIgamma, an isoform of the CaMKI family with an extended C-terminal domain ending with CAAX motif (where AA is aliphatic



Global Trade Item Number

SKUGTIN
C9744-10UG04061837904592