Skip to Content
Merck

C8536

Z-Ala-Arg-Arg 4-methoxy-β-naphthylamide acetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H46N10O6
CAS Number:
Molecular Weight:
690.79
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (TLC)

Quality Segment

form

powder

solubility

ethanol: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Cl.CC(NC(=O)OCc1ccccc1)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(=O)Nc2ccc3ccccc3c2

InChI

1S/C33H44N10O5.ClH/c1-21(40-33(47)48-20-22-9-3-2-4-10-22)28(44)42-27(14-8-18-39-32(36)37)30(46)43-26(13-7-17-38-31(34)35)29(45)41-25-16-15-23-11-5-6-12-24(23)19-25;/h2-6,9-12,15-16,19,21,26-27H,7-8,13-14,17-18,20H2,1H3,(H,40,47)(H,41,45)(H,42,44)(H,43,46)(H4,34,35,38)(H4,36,37,39);1H

InChI key

SRSBQDNWDTXDRD-UHFFFAOYSA-N

Gene Information

human ... CTSB(1508)
mouse ... CTSB(13030)
rat ... CTSB(64529)

General description

Substrate for cathepsin B


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library