Skip to Content
Merck
All Photos(1)

Key Documents

C8534

Sigma-Aldrich

Anti-Chorionic Gonadotropin (α+β subunits) (HCG) antibody produced in rabbit

whole antiserum

Synonym(s):

Chorionic Gonadotropin Antibody, HCG Antibody - Anti-Chorionic Gonadotropin (α+β subunits) (HCG) antibody produced in rabbit, Hcg Antibodies, Hcg Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry: 1:100 using human placental tissue
radioimmunoassay: 1:50,000

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Human Chorionic Gonadotropin (HCG) is a hormone detected in the blood and urine during pregnancy. The peak levels of HCG are detected around Week 12 of gestation and diminish during remaining period of pregnancy. HCG is composed of α and β subunits. There are five variants of HCG that have been identified; HCG, hyperglycosylated HCG, sulphated HCG, free β subunit and pituitary HCG. The important functions of HCG include implantation, angiogenesis, progesterone production, regulation of immune cells and cytotrophoblast differentiation and development of placenta
Anti-Chorionic Gonadotropin (α + α subunits) antibody is specific for both α and β subunits of human chorionic gonadotropin.

Immunogen

HCG

Application

Anti-Chorionic Gonadotropin (α + α subunits) antibody may be used for RIA at a minimum working dilution of 1:50,000. For immunohistochemistry, the recommended working dilution is 1:100-1:200. The antibody was also used for immunofluorescence at a working dilution of 1:50 and for ELISA.

Reconstitution

Dilute with buffer: 0.01 M phosphate, pH 7.8, 0.15 M sodium chloride, 0.5% bovine serum albumin and 0.1 sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Zhao et al.
Oncology letters, 16(4), 4271-4278 (2018-09-11)
Hormones and their receptors affect the development process of gastric cancer. Previous studies have revealed that human chorionic gonadotropin (hCG) is expressed in gastric cancer tissue. However, the mechanism by which hCG exerts its effects on gastric cancer cells had
hCG, the wonder of today's science
Cole LA
Reproductive Biology and Endocrinology, 10, doi: 10-doi: 10 (2010)
W Norris et al.
Placenta, 32 Suppl 2, S182-S185 (2011-02-08)
Human chorionic gonadotropin (hCG) is crucial for successful pregnancy. Its many functions include angiogenesis and immune regulation. Despite years of research, the etiology of preeclampsia remains unknown. Marked by insufficient trophoblast invasion and poor spiral artery remodeling, preeclampsia has also
Biological functions of hCG and hCG-related molecules
Cole LA
Reproductive Biology and Endocrinology, 8, doi: 10-doi: 10 (2010)
Anupam Basu et al.
Analytical biochemistry, 366(2), 175-181 (2007-06-02)
The development of a simultaneous multianalyte immunoassay for the detection of progesterone and human chorionic gonadotropin (hCG) in serum is described. In this simultaneous multianalyte assay, two different enzymes, viz. horse radish peroxidase (HRP) and alkaline phosphatase (ALP), were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service