Skip to Content
Merck

Skip To

C8499

Cucurbitacin B hydrate

≥97% (HPLC)

Synonym(s):

(2β,9β,10α,16α,23E)-25-(acetyloxy)-2,16,20-trihydroxy-9-methyl-19-Norlanosta-5,23-diene-3,11,22-trione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C32H46O8 · xH2O
CAS Number:
Molecular Weight:
558.70 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1(C)C([C@@H](O)C[C@]2([H])C1=CC[C@]([C@@](C[C@@H](O)[C@]3([H])[C@@](C)(O)C(/C=C/C(C)(C)OC(C)=O)=O)(C)[C@]3(C)C4)([H])[C@@]2(C)C4=O)=O

InChI

1S/C32H46O8/c1-17(33)40-27(2,3)13-12-23(36)32(9,39)25-21(35)15-29(6)22-11-10-18-19(14-20(34)26(38)28(18,4)5)31(22,8)24(37)16-30(25,29)7/h10,12-13,19-22,25,34-35,39H,11,14-16H2,1-9H3/b13-12+/t19-,20+,21-,22+,25+,29+,30-,31+,32+/m1/s1

InChI key

IXQKXEUSCPEQRD-DKRGWESNSA-N

Application

Cucurbitacin B hydrate has been used:
  • as a signal transducer and activator of transcription 3 (stat3) inhibitor to determine its effect on the expression of human lysosomal acid lipase (hLAL) in myeloid-derived suppressor cells (MDSCs).
  • as an ecdysone receptor (EcR) antagonist injection to lower the levels of 20-hydoxyecdysone (20E) signaling in butterflies.
  • to determine its effect on the cell viability of pancreatic cancer cell lines.

Biochem/physiol Actions

Cucurbitacin B also exhibits anti-cancer activity by inhibiting telomerase and c-Myc in breast cancer. It also exhibits anti- artherosclerotic activity. Cucurbitacin B also possesses anti-inflammatory and anti-microbial activity.
Cucurbitacin B is a triterpenoid constituent of Cucurbitaceae plant species. Cucurbitacin B inhibits proliferation in a wide variety of tumor cell lines (IC50 15-30 nM) by inducing apoptosis and inducing cell cycle arrest at G2/M phase. Although the mechanism of action is unclear, Cucurbitacin B inhibits STAT 3 phosphorylation and expression levels and has been shown to block JAK2 activity. Curcubitacin B also inhibits the transcriptional activity of HIF1a and Nf-KB. Curcubitacin B is structurally similar to the JAK inhibitor Curcubitacin I.
Cucurbitacin B is an inhibitor of the STAT 3 pathway.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the JAKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C4493SML2008D1756
description

≥97% (HPLC)

description

≥95% (HPLC), solid

description

≥98% (HPLC)

description

≥98% (HPLC), powder

form

powder

form

solid

form

powder

form

powder

assay

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

200

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: >5 mg/mL

solubility

H2O: 2 mg/mL, clear

solubility

methanol: 25 mg/mL

color

white to beige

color

white to off-white

color

white to beige

color

off-white


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters