Skip to Content
Merck

C7604

Anti-γ-Tubulin-Cy3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-γ-Tubulin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
CY3 conjugate
Clone:
polyclonal
Application:
IF (d)
Citations:
11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

CY3 conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human, chicken

technique(s)

direct immunofluorescence: 1:200 using methanol/acetone-fixed chicken fibroblats cells or HeLa cells (human).

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

γ-Tubulin (48 kDa) is a widely expressed and highly conserved protein within the microtubule organizing centers (MTOCs) or centrosome in eukaryotic cells.
Tubulin, γ 1 is a protein encoded by the TUBG1 gene in humans. This gene encodes a member of the tubulin superfamily. γ-tubulin interacts with Arp2/3, a complex that nucleates actin filaments and with the Arp2/3 activator WASH. It is found to have relationship with actin filament dynamics.

Immunogen

synthetic peptide corresponding to the N-terminal region of human γ-tubulin (amino acids 38-53, with C-terminally added lysine) conjugated to KLH. This sequence is specific for γ-tubulin and not found in other members of the tubulin family such as α− β−, δ and ε− tubulins. This sequence is identical in mouse and rat γ-tubulin and highly conserved among species (Drosophila, Aspergillus, and yeast γ-tubulin).

Application

Anti-γ- Tubulin-Cy3 antibody produced in rabbit has been used in direct immunofluorescence (IF)and immunofluorescent staining.
Anti-γ-Tubulin-Cy3 antibody produced in rabbit is suitable for direct immunofluorescence at a dilution of 1:200 using methanol/acetone-fixed chicken fibroblats cells or HeLa cells (human).

Biochem/physiol Actions

γ-Tubulin binds the microtubule minus ends and is responsible for mediating the link between microtubules and the centrosome.
γ-Tubulins in the γ-TuRC might promote microtubule nucleation by providing a template that enhances the intrinsically weak lateral interaction between αβ-tubulin heterodimers when associated laterally. It has a unique functional property of a monomeric tubulin. γ-tubulin is also found to be expressed in lung cancer. Cep70 as an important centrosomal protein that interacts with γ-tubulin and plays a critical role in the regulation of mitotic spindle assembly.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Legal Information

Cy is distributed under license from Amersham Biosciences Limited.
Cy3 is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library