Skip to Content
Merck

C7431

Worthington Accessories

cap/necktube core for 25 L Dewar

Synonym(s):

D025-9C16, cryoware cap, dewar cap, dewar closure, dewar seal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.43
UNSPSC Code:
41121800
Sterility:
non-sterile
Packaging:
pack of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Worthington D025-9C16

technique(s)

cryopreservation: suitable

General description

Replacement necktube core for the LD25 Liquid Dewar.


Still not finding the right product?

Explore all of our products under Worthington Accessories




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Kagistia Hana Utami et al.
Molecular autism, 11(1), 41-41 (2020-05-29)
FXS is the most common genetic cause of intellectual (ID) and autism spectrum disorders (ASD). FXS is caused by loss of FMRP, an RNA-binding protein involved in the translational regulation of a large number of neuronal mRNAs. Absence of FMRP



Global Trade Item Number

SKUGTIN
C7431-1EA04061833574959