Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , plastic (Permanox)
Sterility:
sterile
Packaging:
pack of 16, case of 96
W × l:
25 mm × 75 mm
Wells:
4
material
colorless , plastic (Permanox)
sterility
sterile
feature
CE marked
packaging
pack of 16, case of 96
manufacturer/tradename
Nunc 177437
W × L
25 mm × 75 mm
well area
1.8 cm2
wells
4
working volume
0.5-0.9 mL
suitability
suitable for (viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology)
General description
Grow, stain, and examine cells directly on the Nunc® Lab-Tek® Chamber Slide™, which consists of a polystyrene media chamber attached to a microscopic slide. The slide surface is specially treated for adherent cell culture. Choose either a specialty washed soda lime glass slide or cell attachment optimized Permanox® plastic slide. Media chamber is easily removable without the need for a tool.
Application
- Microscopy of in situ tissue culture
- Viral and mycoplasma testing
- Chromosome studies
- Toxicity testing
- Immunocytology
Features and Benefits
- Cells grow on a standard microscope slide
- Growth chamber can be removed when culturing is complete
- Certified surface treatment ensures optimal contitions for cell attachment and growth
- No cell transfer needed prior to visualisation/staining
- Fits standard equipment
- Saves time and reagents
- Permanox™ does not interfere with fluorescent labels
Legal Information
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Permanox is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nunc® Lab-Tek® Chamber Slide™ system
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.