Skip to Content
Merck
All Photos(1)

Key Documents

C6737

Sigma-Aldrich

Chondroitin sulfate sodium salt from bovine cartilage

98-102% (CPC, titration)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

bovine cartilage

Quality Level

grade

standard (for CPC (cetylpyridinium chloride) titration)

Assay

98-102% (CPC, titration)

form

powder

optical activity

[α]20/D -30 to -20 °, c = 5% (w/v) in water

impurities

<10.0% water (Karl Fischer)

color

white to off-white

solubility

H2O: soluble 50 mg/mL, clear to hazy, colorless to faintly yellow

cation traces

Na: 7.4-8.8% (anhydrous)

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)N[C@H]1[C@H](O)O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@@H]1O[C@H]2O[C@@H]([C@H](O)[C@@H](O)[C@@H]2O)C(O)=O

InChI

1S/C14H23NO15S.Na/c1-3(17)15-5-10(9(30-31(24,25)26)4(2-16)27-13(5)23)28-14-8(20)6(18)7(19)11(29-14)12(21)22;/h4-11,13-14,16,18-20,23H,2H2,1H3,(H,15,17)(H,21,22)(H,24,25,26);/q;+1/p-1/t4-,5-,6-,7-,8+,9+,10-,11+,13-,14+;/m1./s1

InChI key

SHQAYVXGAAXVHV-GNAIRZFJSA-M

General description

Chondroitin, a glucosaminoglycan (GAG), is a polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA) that can be sulfated on the either or both GalNAc and GlcA units. Chondroitin and its sulfates are frequently attached to proteins to form proteoglycans.

Application

Chondroitin sulfate, a glucosaminoglycan (GAG) polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA), is used in a wide variety of areas of research including bioengineering. Chondroitin sulfate may be used as a biomaterial copolymer or surface derivatization reagent in the development of drug delivery vehicles, tissue engineering devices and bioscaffolds. Chondroitin sulfate may be used for the development of biocompatible structures such as hydrogels, sponges, biofilms, microspheres and micelles.

Biochem/physiol Actions

As a large, negatively charged, hydrophilic molecule that is too inflexible to fold into a compact globular structure, chondroitin sulfate molecules form porous hydrated gels, providing mechanical support to tissue and allowing rapid diffusion of water-soluble molecules and migration of cells.

Components

Chondroitin is an acidic mucopolysaccharide found in cartilage, skin, cornea, and umbilical cord. It is composed of alternating N-acetylgalactosamine, D-glucuronic acid, and sulfate residues in equimolar quantities that can be sulfated on either of its subunits. Sulfation can occur at the 6 and/or 4-position of the galactosamine moiety, with potential for any individual disaccharide unit to be 6-sulfated, 4-sulfated, 4,6-disulfated, or unsulfated.

Caution

Store this product at 2-8°C.

Preparation Note

This product is soluble in water at 100 mg/mL.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the chondroitin sulfate disaccharides in dog and horse plasma by HPLC using chondroitinase digestion, precolumn derivatization, and fluorescence detection.
Du J, Eddington N.
Analytical Biochemistry, 306, 252-258 (202)
Feng Wang et al.
Acta biomaterialia, 6(6), 1978-1991 (2009-12-17)
A family of injectable, rapid gelling and highly flexible hydrogel composites capable of releasing insulin-like growth factor (IGF-1) and delivering mesenchymal stromal cell (MSC) were developed. Hydrogel composites were fabricated from Type I collagen, chondroitin sulfate (CS) and a thermosensitive
Satoshi Uchida et al.
Journal of controlled release : official journal of the Controlled Release Society, 155(2), 296-302 (2011-05-17)
Nonviral polycation-based gene carriers (polyplexes) have attracted attention as safe and efficient gene delivery systems. Polyplex micelles comprised of poly(ethyleneglycol)-block-poly{N'-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (PEG-PAsp(DET)) and plasmid DNA (pDNA) have shown high transfection efficiency with low toxicity due to the pH-sensitive protonation behavior of
Zhongming Liang et al.
Journal of pharmaceutical and biomedical analysis, 28(2), 245-249 (2002-04-04)
A photometric titration method was developed and validated to quantitate sodium chondroitin sulfate in raw materials and Cosequin DS chewable tablet(1). About 0.1% (w/v) cetylpyridinium chloride was used to titrate sodium chondroitin sulfate with photometric indication at wavelength 420 nm.
Maelene L Wong et al.
Acta biomaterialia, 9(5), 6492-6501 (2013-01-17)
The ability of residual antigens on decellularized tissue to elicit the immune response upon implantation motivates development of a more rigorous antigen removal (AR) process for xenogeneic scaffold generation. Antigen removal strategies promoting solubilization of hydrophilic proteins (predominantly cytoplasmic) enhance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service