Skip to Content
Merck

C6442

Nalgene® rectangular carboy

nominal capacity 20 L (5 gal), Style 2211, autoclavable: no

Synonym(s):

carboys, nalgene carboys, Nalgene 2211-0050, laboratory container, receptical, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41121820
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

high-density polyethylene Carboy, polypropylene screw closure

feature

autoclavable: no

packaging

pack of 1 ea

manufacturer/tradename

Nalgene 2211-0050

L × W × H

12 1/2 in. (318 mm) × 8 5/8 in. (219 mm) × 15 in. (381 mm)

nominal capacity

20 L (5 gal)

General description

Molded-in graduations in liters and gallons. Built-in shoulder loops with a stainless steel bail handle attached. Wide-mouth opening (100 mm)

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nalgene® rectangular carboy




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library