About This Item
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , polystyrene chamber
Sterility:
sterile
Packaging:
pack of 16 ea, case of 96 ea
Wells:
1
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
packaging
pack of 16 ea, case of 96 ea
manufacturer/tradename
Nunc 154453
technique(s)
cell culture: suitable
well area
8.6 cm2
wells
1
working volume
2.0-4.5 mL
suitability
suitable for (cell culture; attachment and growth of cells)
General description
Grow, stain, and examine cells directly on the Nunc® Lab-Tek® II chamber slide™ which consists of a removable polystyrene media chamber attached to a standard glass slide. Slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features and Benefits
- Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
- Tabbed polystyrene lid allows gas exchange with minimal evaporation.
- Removal tool separates media chamber from glass slide.
- Biocompatible adhesive hydrophobic well border is printed on slide.
- Slide features a white frosted writing area.
- Includes: Chamber removal tool.
- Sterile
- Packed in convenient stackable incubation trays.
Legal Information
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nunc® Lab-Tek® II Chamber Slide™ system
Choose from one of the most recent versions:
Already Own This Product?