Skip to Content
Merck
All Photos(3)

Key Documents

C6022

Sigma-Aldrich

Cyproheptadine hydrochloride sesquihydrate

≥98% (TLC), solid

Synonym(s):

Piperidine, 4-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-, hydrochloride, hydrate (2:3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N·HCl·1.5H2O
CAS Number:
Molecular Weight:
350.88
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

solid

color

white to slightly yellow

solubility

ethanol: soluble
methanol: soluble

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

SMILES string

Cl.CN1CCC(\CC1)=C2/c3ccccc3C=Cc4ccccc24

InChI

1S/2C21H21N.2ClH.3H2O/c2*1-22-14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21;;;;;/h2*2-11H,12-15H2,1H3;2*1H;3*1H2

InChI key

ZEAUHIZSRUAMQG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Cyproheptadine hydrochloride sesquihydrate has been used in:
  • testing anti-inflammatory activity in serotonin receptor (5-HT) induced inflammation[1]
  • the inhibition of 5-HT in in vivo and in vitro bioassays in crabs[2]
  • the inhibition of 5-HT in embryo physiological experiments[3]

inhibition of calcitonin gene related peptide (CGRP)[4]

Biochem/physiol Actions

Cyproheptadine hydrochloride sesquihydrate is a serotonin receptor (5-HT2/5-HT1C) antagonist,[5] H1 histamine receptor antagonist[6] and an antipruritic.[7] The inhibition of 5-HT by cyproheptadine improves cognitive function in schizophrenia disorder.[5] Cyproheptadine is effective for treating functional gastrointestinal disorders (FGIDs).[8] Food and Drug Administration (FDA) approved cyproheptadine, has antidepressant and antiplatelet functionality. It may be effective in treating thromboembolic disorders.[9] Cyproheptadine inhibits lysine methyltransferase 7/9 (Set7/9) leading to a decrease in estrogen receptor (ERα) expression and growth arrest in breast cancer cells.[10]

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Observations on the use of cyproheptadine hydrochloride as an antipruritic agent in allergic cats.
Scott DW, et al.
The Canadian Veterinary Journal. La Revue Veterinaire Canadienne, 39(10), 634-634 (1998)
The analgesic agent tapentadol inhibits calcitonin gene-related peptide release from isolated rat brainstem via a serotonergic mechanism
Greco MC, et al.
Life Sciences, 145, 161-165 (2016)
The antidepressant 5-HT2A receptor antagonists pizotifen and cyproheptadine inhibit serotonin-enhanced platelet function
Lin OA, et al.
PLoS ONE, 9(1), e87026-e87026 (2014)
Comparative anticholinergic activities of 10 histamine H1 receptor antagonists in two functional models
Orzechowski RF, et al.
European Journal of Pharmacology, 506(3), 257-264 (2005)
H W Suh et al.
Neuropeptides, 33(2), 121-129 (2000-02-05)
The present study was designed to investigate the modulatory effects of blockade of spinal histamine receptors on antinociception induced by supraspinally administered mu-epsilon-, delta-, and kappa-opioid receptor agonists. The effects of intrathecal (i.t.) injections with cyproheptadine [a histamine-1 (H1) receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service