Skip to Content
Merck

Skip To

C5894

Collagenase from Clostridium histolyticum

non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid

Synonym(s):

Clostridiopeptidase A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-582-9
MDL number:
EC Number:
Specific activity:
≥125 CDU/mg solid, 0.5-5.0 FALGPA units/mg solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile; 0.2 μm filtered

form

lyophilized powder

specific activity

≥125 CDU/mg solid, 0.5-5.0 FALGPA units/mg solid

mol wt

68-130 kDa

storage temp.

−20°C

Application

Collagenase has been used in the preparation of arterial tissue for the study of Advanced Glycosylation End Products (AGE). The enzyme has also been used along with other proteases for the disaggregation of human tumor, mouse kidney, human brain, lung epithelium and many other tissues. It is also used for liver and kidney perfusion studies, digestion of pancreas, and isolation of nonparenchymal hepatocytes. This enzyme is tested for the release of hepatocytes at approx.1 mg/mL conc. (in a total volume of 100 mL for each rat liver).

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. The pH optimum is 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Collagenase treatment can cause some cells to die. Typically, concentrations varying from 0.1 to 5 mg/mL are used for digestion. The duration of reaction varyies from 15 minutes to several hours and yields a satisfactory cell dissociation without causing too much cell death. Krebs Ringer buffer with calcium and BSA is preferred and Zn2+ is required for activity.

Preparation Note

Solutions are typically prepared at 1-2 mg/mL in TESCA buffer (containing 50 mM TES, 0.36 mM Calcium chloride, pH 7.4 at 37°C.

Other Notes

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Disclaimer

As supplied, this product is stable for one year at -20°C. There is no loss in FALGPA or protease activity in 30 days at 37°C, 50°C and -20°C. Solutions of crude collagenase are stable if frozen quickly in aliquots (at 10 mg/mL) and kept frozen at -20°C. Further freeze-thaw cycles will damage the solution. The product retains 100% activity over 7 hours when held on ice.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2014C1639C7657
specific activity

≥125 CDU/mg solid, 0.5-5.0 FALGPA units/mg solid

specific activity

≥1 FALGPA units/mg solid, ≥125 CDU/mg solid

specific activity

≥125 CDU/mg solid, 0.2-1.0 FALGPA units/mg solid

specific activity

≥800 CDU/mg solid, 2-5 FALGPA units/mg solid

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

powder

sterility

non-sterile; 0.2 μm filtered

sterility

0.2 μm filtered

sterility

0.2 μm filtered

sterility

-

mol wt

68-130 kDa

mol wt

68-125 kDa

mol wt

68-130 kDa

mol wt

68-130 kDa

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters