Skip to Content
Merck
All Photos(1)

Key Documents

C5429

Sigma-Aldrich

Z-Arg-Arg-7-amido-4-methylcoumarin hydrochloride

Synonym(s):

Z-RR-AMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H39N9O6
CAS Number:
Molecular Weight:
621.69
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

form

powder

impurities

~4% isopropanol

storage temp.

−20°C

SMILES string

Cl.CC1=CC(=O)Oc2cc(NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)OCc3ccccc3)ccc12

InChI

1S/C30H39N9O6.ClH/c1-18-15-25(40)45-24-16-20(11-12-21(18)24)37-26(41)22(9-5-13-35-28(31)32)38-27(42)23(10-6-14-36-29(33)34)39-30(43)44-17-19-7-3-2-4-8-19;/h2-4,7-8,11-12,15-16,22-23H,5-6,9-10,13-14,17H2,1H3,(H,37,41)(H,38,42)(H,39,43)(H4,31,32,35)(H4,33,34,36);1H

InChI key

HLSNWWKWVKPXKI-UHFFFAOYSA-N

Gene Information

human ... CTSB(1508)
mouse ... CTSB(13030)
rat ... CTSB(64529)

Amino Acid Sequence

Z-Arg-Arg-AMC

Application

Z-Arg-Arg-7-amido-4-methylcoumarin hydrochloride has been used:
  • as a cathepsin B substrate in zymography analysis of protease in feces extract.
  • in cathepsin B activity assay.
  • as a fluorogenic peptide substrate to determine the matrix metalloproteinase enzyme activity.

Substrates

Fluorogenic substrate for cathepsin B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomas Erban et al.
Frontiers in physiology, 7, 53-53 (2016-03-05)
Tyrophagus putrescentiae (Schrank, 1781) is an emerging source of allergens in stored products and homes. Feces proteases are the major allergens of astigmatid mites (Acari: Acaridida). In addition, the mites are carriers of microorganisms and microbial adjuvant compounds that stimulate
High pressure effects on the activities of cathepsins B and D of mackerel and horse mackerel muscle
Fidalgo L G
Czech Journal of Food Sciences, 7, 53-53 (2014)
Kai S Beckwith et al.
Nature communications, 11(1), 2270-2270 (2020-05-10)
Mycobacterium tuberculosis is a global health problem in part as a result of extensive cytotoxicity caused by the infection. Here, we show how M. tuberculosis causes caspase-1/NLRP3/gasdermin D-mediated pyroptosis of human monocytes and macrophages. A type VII secretion system (ESX-1)
Greta Geldenhuys et al.
Journal of the science of food and agriculture, 96(1), 271-279 (2015-02-03)
Baseline research on the toughness of Egyptian goose meat is required. This study therefore investigates the post mortem pH and temperature decline (15 min-4 h 15 min post mortem) in the pectoralis muscle (breast portion) of this gamebird species. It
Miguel G Ximénez-Embún et al.
PloS one, 11(1), e0145275-e0145275 (2016-01-07)
Climate change will bring more drought periods that will have an impact on the irrigation practices of some crops like tomato, from standard water regime to deficit irrigation. This will promote changes in plant metabolism and alter their interactions with

Articles

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service