Skip to Content
Merck

Skip To

C4805

β-Cyclodextrin

powder, BioReagent, suitable for cell culture, ≥97%

Synonym(s):

Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
UNSPSC Code:
12352201
NACRES:
NA.77
PubChem Substance ID:
EC Number:
231-493-2
Beilstein/REAXYS Number:
78623
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

product line

BioReagent

assay

≥97%

form

powder

optical activity

[α]20/D +162±3°, c = 1.5% in H2O

technique(s)

cell culture | mammalian: suitable

mp

290-300 °C (dec.) (lit.)

solubility

1 M NaOH: 50 mg/mL

application(s)

advanced drug delivery

shipped in

ambient

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

Application

β-Cyclodextrin(β-CD) has been used in the synthesis of mono-6-tosyl-CD. It has also been used as a constituent in the serum-free medium to assess the involvement of cholesterol-dependent mechanisms in the uptake of nanoparticles by rat basophilic leukemia cells.

Biochem/physiol Actions

β-Cyclodextrin is the cyclic α heptamer of glucose. It acts as a host to form inclusion compounds with the guests including derivatives of benzene, cyclohexane, adamantane, other alicyclic guests, and also inorganic molecules or ions. It is generally used to solubilize non-polar compounds such a fatty acids, lipids and cholesterol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W402826C4930C4680
technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

powder

form

powder

form

powder

form

powder

application(s)

advanced drug delivery

application(s)

flavors and fragrances

application(s)

-

application(s)

-

sterility

non-sterile

sterility

-

sterility

-

sterility

non-sterile

solubility

1 M NaOH: 50 mg/mL

solubility

-

solubility

1 M NaOH: 25 mg/mL, clear to slightly hazy

solubility

H2O: 10 mg/mL

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage temp.

room temp


Still not finding the right product?

Explore all of our products under β-Cyclodextrin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters