Skip to Content

C-41330

Macrophage Detachment Solution DXF

250 ml

Synonym(s):

Macrophage Detachment Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

PromoCell®

form

aqueous solution

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Macrophage Detachment Solution DXF

Application

Macrophages are strongly adherent and are not effectively dislodged from culture surfaces by standard enzymatic cell dissociation reagents. Additionally, these cells are highly sensitive to improper dissociation procedures. The Macrophage Detachment Solution DXF is non-enzymatic, defined and animal component-free. It was especially designed for the gentle release of adherent macrophages and guarantees the best possible cell viability, even after prolonged exposure times. Unlike enzyme-based solutions, the Macrophage Detachment Solution DXF does not alter cell surface proteins and neutralization is not necessary.Note: The Macrophage Detachment Solution DXF is not suitable for the dissociation of other adherent cell types.

Preparation Note

Click here for more information.
Store at 2 to 8°C immediately after arrival. If stored properly, the product is stable until the expiry date stated on the label.

Analysis Note

All lots of PromoCell Macrophage Detachment Solution DXF are subjected to comprehensive quality control tests. Each lot is routinely tested for biological function and absence of cytotoxicity. Approved in-house lots are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Legal Information

PromoCell is a registered trademark of PromoCell GmbH


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library