Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.71
UNSPSC Code:
12352207
Pricing and availability is not currently available.
packaging
pkg of 3 × 250 ml
technique(s)
cell culture | mammalian: suitable
shipped in
cooler
storage temp.
−20°C
General description
DetachKit-250
Application
The DetachKit was developed for gentle and effective detachment of adherent primary human cells in culture. Each batch is tested on primary human cells. The DetachKit is optimized for primary human cells but can also be used for primary animal cells and cell lines.
Preparation Note
Click here for more information.
Store at -20°C in the dark immediately after arrival. Refrigerate thawed solution at 4 to 8°C. If stored properly, the product is stable until the expiry date stated on the label. Thawed solutions can be stored at 4 to 8°C and used for up to 6 weeks.
Analysis Note
All lots of PromoCell′s DetachKits are subjected to comprehensive quality control tests. Each lot is routinely tested for use with primary human cells, the absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Other Notes
Each DetachKit consists of three components: HEPES BSS (HEPES buffered Balanced Salt Solution), Trypsin/EDTA Solution and TNS (Trypsin Neutralization Solution). The DetachKit comes with a Trypsin/EDTA ratio of 0.04%/0.03%. HEPES BSS contains 30 mM HEPES, D-Glucose, NaCl, KCl, Na-Phosphate and Phenol Red. TNS contains 0.05% Trypsin Inhibitor from soybean and 0.1% Bovine Serum Albumin.
Still not finding the right product?
Explore all of our products under DetachKit
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.