Skip to Content
Merck

C4018

Choloylglycine Hydrolase from Clostridium perfringens (C. welchii)

lyophilized powder, ≥100 units/mg protein

Synonym(s):

3α,7α,12α-Trihydroxy-5β-cholan-24-oylglycine amidohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥100 units/mg protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

specific activity

≥100 units/mg protein

composition

Protein, ≥50% biuret

shipped in

wet ice

storage temp.

−20°C

Application

The enzyme from Sigma has been used in the analysis of bile samples in various studies.

Biochem/physiol Actions

The enzyme is an N-terminal nucleophilic (Ntn) hydrolase that catalyzes the hydrolysis of amide bonds, liberates the glycine/taurine moiety from the steroid core and eventually yields unconjugated bile acids. Agents that oxidize thiol groups (e.g., p-mercuribenzoate, iodoacetamide, Hg2+, Cu2+, and Cd2+) have been shown to strongly inhibit bile salt hydrolase (BSH) activity in Clostridium perfringens.

Physical form

Partially purified lyophilized powder containing buffer salts and stabilizer

Other Notes

One unit will hydrolyze 1.0 μmole of glycocholic acid to glycine and cholic acid in 5 min at pH 5.6 at 37 °C.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library